Learn More
Hampton Research HR2-907-04/SaltRx 1 #04 (A4), 185 ml - 1.5 M Ammonium chloride, 0.1 M BIS-TRIS propane pH 7.0
SDP
Catalog No. 501221
Encompass
HR2-907-04/SaltRx 1 #04 (A4), 185 ml - 1.5 M Ammonium chloride, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-122-1 Supplier Hampton Research Supplier No. HR290704
Add to Cart
Add to Cart
HR2-907-04/SaltRx 1 #04 (A4), 185 ml - 1.5 M Ammonium chloride, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.