Learn More
Hampton Research HR2-907-07/SaltRx 1 #07 (A7), 185 ml - 3.5 M Ammonium chloride, 0.1 M BIS-TRIS propane pH 7.0
SDP
Catalog No. 501224
Encompass
HR2-907-07/SaltRx 1 #07 (A7), 185 ml - 3.5 M Ammonium chloride, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-122-4 Supplier Hampton Research Supplier No. HR290707
HR2-907-07/SaltRx 1 #07 (A7), 185 ml - 3.5 M Ammonium chloride, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.