Learn More
Hampton Research HR2-907-08/SaltRx 1 #08 (A8), 185 ml - 3.5 M Ammonium chloride, 0.1 M Tris pH 8.5
SDP
Catalog No. 501225
Encompass
HR2-907-08/SaltRx 1 #08 (A8), 185 ml - 3.5 M Ammonium chloride, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.
Catalog No. 50-122-5 Supplier Hampton Research Supplier No. HR290708
HR2-907-08/SaltRx 1 #08 (A8), 185 ml - 3.5 M Ammonium chloride, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.