Learn More
Hampton Research HR2-907-23/SaltRx 1 #23 (B11), 185 ml - 0.4 M Magnesium formate dihydrate, 0.1 M Sodium acetate trihydrate pH 4.6
SDP
Catalog No. 501240
Encompass
HR2-907-23/SaltRx 1 #23 (B11), 185 ml - 0.4 M Magnesium formate dihydrate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-124-0 Supplier Hampton Research Supplier No. HR290723
HR2-907-23/SaltRx 1 #23 (B11), 185 ml - 0.4 M Magnesium formate dihydrate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.