Learn More
Hampton Research HR2-907-27/SaltRx 1 #27 (C3), 185 ml - 2.0 M Sodium formate, 0.1 M Sodium acetate trihydrate pH 4.6
SDP
Catalog No. 501244
Encompass
HR2-907-27/SaltRx 1 #27 (C3), 185 ml - 2.0 M Sodium formate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-124-4 Supplier Hampton Research Supplier No. HR290727
HR2-907-27/SaltRx 1 #27 (C3), 185 ml - 2.0 M Sodium formate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.