Learn More
Hampton Research HR2-907-31/SaltRx 1 #31 (C7), 185 ml - 3.5 M Sodium formate, 0.1 M BIS-TRIS propane pH 7.0
SDP
Catalog No. 501248
Encompass
HR2-907-31/SaltRx 1 #31 (C7), 185 ml - 3.5 M Sodium formate, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-124-8 Supplier Hampton Research Supplier No. HR290731
Add to Cart
Add to Cart
HR2-907-31/SaltRx 1 #31 (C7), 185 ml - 3.5 M Sodium formate, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.