Learn More
Hampton Research HR2-907-34/SaltRx 1 #34 (C10), 185 ml - 2.2 M DL-Malic acid pH 7.0, 0.1 M BIS-TRIS propane pH 7.0
SDP
Catalog No. 501251
Encompass
HR2-907-34/SaltRx 1 #34 (C10), 185 ml - 2.2 M DL-Malic acid pH 7.0, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-125-1 Supplier Hampton Research Supplier No. HR290734
HR2-907-34/SaltRx 1 #34 (C10), 185 ml - 2.2 M DL-Malic acid pH 7.0, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.