Learn More
Hampton Research HR2-907-36/SaltRx 1 #36 (C12), 185 ml - 2.4 M Sodium malonate pH 7.0, 0.1 M BIS-TRIS propane pH 7.0
Catalog No. 501253
Encompass
HR2-907-36/SaltRx 1 #36 (C12), 185 ml - 2.4 M Sodium malonate pH 7.0, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-125-3 Supplier Hampton Research Supplier No. HR290736
HR2-907-36/SaltRx 1 #36 (C12), 185 ml - 2.4 M Sodium malonate pH 7.0, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.