Learn More
Hampton Research HR2-907-39/SaltRx 1 #39 (D3), 185 ml - 2.5 M Ammonium nitrate, 0.1 M Tris pH 8.5
SDP
Catalog No. 501256
Encompass
HR2-907-39/SaltRx 1 #39 (D3), 185 ml - 2.5 M Ammonium nitrate, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.
Catalog No. 50-125-6 Supplier Hampton Research Supplier No. HR290739
HR2-907-39/SaltRx 1 #39 (D3), 185 ml - 2.5 M Ammonium nitrate, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.