Learn More
Hampton Research HR2-907-41/SaltRx 1 #41 (D5), 185 ml - 6.0 M Ammonium nitrate, 0.1 M BIS-TRIS propane pH 7.0
SDP
Catalog No. 501258
Encompass
HR2-907-41/SaltRx 1 #41 (D5), 185 ml - 6.0 M Ammonium nitrate, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-125-8 Supplier Hampton Research Supplier No. HR290741
HR2-907-41/SaltRx 1 #41 (D5), 185 ml - 6.0 M Ammonium nitrate, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.