Learn More
Hampton Research HR2-907-43/SaltRx 1 #43 (D7), 185 ml - 1.5 M Sodium nitrate, 0.1 M Sodium acetate trihydrate pH 4.6
SDP
Catalog No. 501260
Encompass
HR2-907-43/SaltRx 1 #43 (D7), 185 ml - 1.5 M Sodium nitrate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-126-0 Supplier Hampton Research Supplier No. HR290743
HR2-907-43/SaltRx 1 #43 (D7), 185 ml - 1.5 M Sodium nitrate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.