Learn More
Hampton Research HR2-907-46/SaltRx 1 #46 (D10), 185 ml - 4.0 M Sodium nitrate, 0.1 M Sodium acetate trihydrate pH 4.6
SDP
Catalog No. 501263
Encompass
HR2-907-46/SaltRx 1 #46 (D10), 185 ml - 4.0 M Sodium nitrate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-126-3 Supplier Hampton Research Supplier No. HR290746
HR2-907-46/SaltRx 1 #46 (D10), 185 ml - 4.0 M Sodium nitrate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.