Learn More
Hampton Research HR2-909-01/SaltRx 2 #01 (E01), 185 ml - 1.0 M Ammonium phosphate monobasic, 0.1 M Sodium acetate trihydrate pH 4.6
SDP
Catalog No. 501362
Encompass
HR2-909-01/SaltRx 2 #01 (E01), 185 ml - 1.0 M Ammonium phosphate monobasic, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-136-2 Supplier Hampton Research Supplier No. HR290901
HR2-909-01/SaltRx 2 #01 (E01), 185 ml - 1.0 M Ammonium phosphate monobasic, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.