Learn More
Hampton Research HR2-909-02/SaltRx 2 #02 (E02), 185 ml - 1.8 M Ammonium phosphate monobasic, 0.1 M Sodium acetate trihydrate pH 4.6
SDP
Catalog No. 501363
Encompass
HR2-909-02/SaltRx 2 #02 (E02), 185 ml - 1.8 M Ammonium phosphate monobasic, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-136-3 Supplier Hampton Research Supplier No. HR290902
HR2-909-02/SaltRx 2 #02 (E02), 185 ml - 1.8 M Ammonium phosphate monobasic, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.