Learn More
Hampton Research HR2-909-13/SaltRx 2 #13 (F01), 185 ml - 1.5 M Ammonium sulfate, 0.1 M Sodium acetate trihydrate pH 4.6
SDP
Catalog No. 501374
Encompass
HR2-909-13/SaltRx 2 #13 (F01), 185 ml - 1.5 M Ammonium sulfate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-137-4 Supplier Hampton Research Supplier No. HR290913
Add to Cart
Add to Cart
HR2-909-13/SaltRx 2 #13 (F01), 185 ml - 1.5 M Ammonium sulfate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.