Learn More
Hampton Research SALTRX 2 NO14 185ML
SDP
Catalog No. 501375
Encompass
HR2-909-14/SaltRx 2 #14 (F02), 185 ml - 1.5 M Ammonium sulfate, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-137-5 Supplier Hampton Research Supplier No. HR290914
HR2-909-14/SaltRx 2 #14 (F02), 185 ml - 1.5 M Ammonium sulfate, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.