Learn More
Hampton Research SALTRX 2 NO18 185ML
SDP
Catalog No. 501379
Encompass
HR2-909-18/SaltRx 2 #18 (F06), 185 ml - 2.5 M Ammonium sulfate, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.
Catalog No. 50-137-9 Supplier Hampton Research Supplier No. HR290918
Add to Cart
Add to Cart
HR2-909-18/SaltRx 2 #18 (F06), 185 ml - 2.5 M Ammonium sulfate, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.