Learn More
Hampton Research SALTRX 2 NO19 185ML
SDP
Catalog No. 501380
Encompass
HR2-909-19/SaltRx 2 #19 (F07), 185 ml - 0.8 M Lithium sulfate monohydrate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-138-0 Supplier Hampton Research Supplier No. HR290919
HR2-909-19/SaltRx 2 #19 (F07), 185 ml - 0.8 M Lithium sulfate monohydrate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.