Learn More
Hampton Research SALTRX 2 NO25 185ML
SDP
Supplier:  Hampton Research HR290925
Encompass
HR2-909-25/SaltRx 2 #25 (G01), 185 ml - 1.0 M Magnesium sulfate hydrate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-138-6