Learn More
Hampton Research SALTRX 2 NO26 185ML
SDP
Catalog No. 501387
Encompass
HR2-909-26/SaltRx 2 #26 (G02), 185 ml - 1.0 M Magnesium sulfate hydrate, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-138-7 Supplier Hampton Research Supplier No. HR290926
HR2-909-26/SaltRx 2 #26 (G02), 185 ml - 1.0 M Magnesium sulfate hydrate, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.