Learn More
Hampton Research SALTRX 2 NO28 185ML
SDP
Catalog No. 501389
Encompass
HR2-909-28/SaltRx 2 #28 (G04), 185 ml - 1.8 M Magnesium sulfate hydrate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-138-9 Supplier Hampton Research Supplier No. HR290928
HR2-909-28/SaltRx 2 #28 (G04), 185 ml - 1.8 M Magnesium sulfate hydrate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.