Learn More
Hampton Research SALTRX 2 NO29 185ML
SDP
Catalog No. 501390
Encompass
HR2-909-29/SaltRx 2 #29 (G05), 185 ml - 1.8 M Magnesium sulfate hydrate, 0.1 M BIS-TRIS propane pH 7.0
Catalog No. 50-139-0 Supplier Hampton Research Supplier No. HR290929
Add to Cart
Add to Cart
HR2-909-29/SaltRx 2 #29 (G05), 185 ml - 1.8 M Magnesium sulfate hydrate, 0.1 M BIS-TRIS propane pH 7.0