Learn More
Hampton Research SALTRX 2 NO30 185ML
SDP
Catalog No. 501391
Encompass
HR2-909-30/SaltRx 2 #30 (G06), 185 ml - 1.8 M Magnesium sulfate hydrate, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.
Catalog No. 50-139-1 Supplier Hampton Research Supplier No. HR290930
Add to Cart
Add to Cart
HR2-909-30/SaltRx 2 #30 (G06), 185 ml - 1.8 M Magnesium sulfate hydrate, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.