Learn More
Hampton Research SALTRX 2 NO31 185ML
SDP
Catalog No. 501392
Encompass
HR2-909-31/SaltRx 2 #31 (G07), 185 ml - 0.7 M Ammonium tartrate dibasic, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-139-2 Supplier Hampton Research Supplier No. HR290931
HR2-909-31/SaltRx 2 #31 (G07), 185 ml - 0.7 M Ammonium tartrate dibasic, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.