Learn More
Hampton Research SALTRX 2 NO33 185ML
SDP
Catalog No. 501394
Encompass
HR2-909-33/SaltRx 2 #33 (G09), 185 ml - 0.7 M Ammonium tartrate dibasic, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.
Catalog No. 50-139-4 Supplier Hampton Research Supplier No. HR290933
HR2-909-33/SaltRx 2 #33 (G09), 185 ml - 0.7 M Ammonium tartrate dibasic, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.