Learn More
Hampton Research SALTRX 2 NO34 185ML
SDP
Catalog No. 501395
Encompass
HR2-909-34/SaltRx 2 #34 (G10), 185 ml - 1.0 M Ammonium tartrate dibasic, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-139-5 Supplier Hampton Research Supplier No. HR290934
HR2-909-34/SaltRx 2 #34 (G10), 185 ml - 1.0 M Ammonium tartrate dibasic, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.