Learn More
Hampton Research SALTRX 2 NO35 185ML
SDP
Catalog No. 501396
Encompass
HR2-909-35/SaltRx 2 #35 (G11), 185 ml - 1.3 M Ammonium tartrate dibasic, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-139-6 Supplier Hampton Research Supplier No. HR290935
HR2-909-35/SaltRx 2 #35 (G11), 185 ml - 1.3 M Ammonium tartrate dibasic, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.