Learn More
Hampton Research SALTRX 2 NO36 185ML
SDP
Catalog No. 501397
Encompass
HR2-909-36/SaltRx 2 #36 (G12), 185 ml - 1.4 M Ammonium tartrate dibasic, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.
Catalog No. 50-139-7 Supplier Hampton Research Supplier No. HR290936
HR2-909-36/SaltRx 2 #36 (G12), 185 ml - 1.4 M Ammonium tartrate dibasic, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.