Learn More
Hampton Research SALTRX 2 NO40 185ML
SDP
Catalog No. 501401
Encompass
HR2-909-40/SaltRx 2 #40 (H04), 185 ml - 1.2 M Potassium sodium tartrate tetrahydrate, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.
Catalog No. 50-140-1 Supplier Hampton Research Supplier No. HR290940
HR2-909-40/SaltRx 2 #40 (H04), 185 ml - 1.2 M Potassium sodium tartrate tetrahydrate, 0.1 M Tris pH 8.5. Please note items from this supplier are non-returnable.