Learn More
Hampton Research SALTRX 2 NO41 185ML
SDP
Catalog No. 501402
Encompass
HR2-909-41/SaltRx 2 #41 (H05), 185 ml - 0.5 M Potassium thiocyanate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.
Catalog No. 50-140-2 Supplier Hampton Research Supplier No. HR290941
HR2-909-41/SaltRx 2 #41 (H05), 185 ml - 0.5 M Potassium thiocyanate, 0.1 M Sodium acetate trihydrate pH 4.6. Please note items from this supplier are non-returnable.