Learn More
Hampton Research SALTRX 2 NO45 185ML
SDP
Catalog No. 501406
Encompass
HR2-909-45/SaltRx 2 #45 (H09), 185 ml - 4.0 M Ammonium acetate, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-140-6 Supplier Hampton Research Supplier No. HR290945
HR2-909-45/SaltRx 2 #45 (H09), 185 ml - 4.0 M Ammonium acetate, 0.1 M BIS-TRIS propane pH 7.0. Please note items from this supplier are non-returnable.