Learn More
Apexbio Technology LLC Salubrinal, 405060-95-9, MFCD00548612, 10 mg
SDP
Supplier:  Apexbio Technology LLC B202510
Encompass_Preferred
MF: C21H17Cl3N4OS, MW: 479.81, Solubility: 48mg/mL in DMSO. A cell-permeable and selective inhibitor of eIF2alpha dephosphorylation with an IC50 of 15 M.
Catalog No. 50-101-1234