Learn More
Medchemexpress LLC HY-15486 10mg Medchemexpress, Salubrinal CAS:405060-95-9 Purity:>98%
SDP
Catalog No. 502029129
Encompass_Preferred
Medchemexpress, HY-15486 10mg Salubrinal CAS:405060-95-9 Salubrinal is a cell-permeable and selective inhibitor of eIF2α dephosphorylation. Salubrinal acts as a dual-specificity phosphatase 2 (Dusp2) inhibitor and suppresses inflammation in anti-collagen antibody-induced arthritis[2]. Salubrinal has antiviral activity against HSV-1 and inhibits dephosphorylation of eIF2α mediated by the HSV-1 protein ICP34.5[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9129 Supplier Medchemexpress LLC Supplier No. HY1548610MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-15486 10mg Salubrinal CAS:405060-95-9 Salubrinal is a cell-permeable and selective inhibitor of eIF2α dephosphorylation. Salubrinal acts as a dual-specificity phosphatase 2 (Dusp2) inhibitor and suppresses inflammation in anti-collagen antibody-induced arthritis[2]. Salubrinal has antiviral activity against HSV-1 and inhibits dephosphorylation of eIF2α mediated by the HSV-1 protein ICP34.5[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.