Learn More
TARGETMOL CHEMICALS INC SALUBRINAL 25MG
SDP
Catalog No. 502580415
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Salubrinal a phosphatases (PP1) inhibitor(IC50=1.7 uM) exhibits function on the eukaryotic translation initiation factor 2 subunit (eIF2(alpha)). purity: 99%
Catalog No. 50-258-0415 Supplier TARGETMOL CHEMICALS INC Supplier No. T304525MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Salubrinal a phosphatases (PP1) inhibitor(IC50=1.7 uM) exhibits function on the eukaryotic translation initiation factor 2 subunit (eIF2(alpha)). purity: 99%