Learn More
Enzo Life Sciences Salubrinal (5mg). CAS: 405060-95-9
SDP
Catalog No. 502008552
Encompass_Preferred
Cell permeable and selective inhibitor of the phosphatase complexes that dephosphorylate eukaryotic translation initiation factor 2 subunit α (eIF-2α). Protects ER stress-induced apoptosis (EC50~15µM in PC12 cells stimulated with 750ng/ml of tunicamycin (BML-CC104)). Purity: ≥98% (HPLC(UV)). Appearance: White to off-white powder. Solubility: Soluble in DMSO (300 mg/ml), dioxane (0.5 mg/ml), methanol (5 mg/ml) or acetonitrile (1.5 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8552 Supplier Enzo Life Sciences Supplier No. ALX270428M005
Cell permeable and selective inhibitor of the phosphatase complexes that dephosphorylate eukaryotic translation initiation factor 2 subunit α (eIF-2α). Protects ER stress-induced apoptosis (EC50~15µM in PC12 cells stimulated with 750ng/ml of tunicamycin (BML-CC104)). Purity: ≥98% (HPLC(UV)). Appearance: White to off-white powder. Solubility: Soluble in DMSO (300 mg/ml), dioxane (0.5 mg/ml), methanol (5 mg/ml) or acetonitrile (1.5 mg/ml). Long Term Storage: -20°C.