Learn More
Bullard™ SALUS HC Powered Air Purifying Respirator System
SDP
Catalog No. 19087985 Shop All Bullard Company Products
Change view
Click to view available options
Includes:
Narrow LF Hood, Shoulder Carriage, Blower Motor, 2 HEPA Filters, 2 Batteries, and Battery Charger
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Size
19-087-985 Narrow LF Hood, Shoulder Carriage, Blower Motor, 2 HEPA Filters, 2 Batteries, and Battery Charger Medium
19-087-984 Narrow LF Hood, Shoulder Carriage, Blower Motor, 2 HEPA Filters, 2 Batteries, and Battery Charger Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-087-985 Supplier Bullard™ Supplier No. SALHC20LF2M2
SALUS HC high performing respiratory solution is designed to protect clinicians from exposure to particulates and biological aerosols including viruses and bacteria. Choose from two different sizes and systems that offer one or two HEPA filters and batteries.
  • Designed for healthcare and EMS professionals
  • New NIOSH PAPR 100-P standard
  • Unique shoulder carriage design
  • Compact, lightweight, comfortable
  • Two speeds (low/high)
Includes:
  • Medium or Large Narrow LF Hood
  • Blower Motor
  • Shoulder Carriage
  • HEPA Filter - 1 or 2
  • Battery - 1 or 2
  • Battery Charger
Specifications
Assigned Protection Factor (APF) 25
Battery Life 9.5 hours
Battery Type Lithium-ion
Breathing Tube Length 7.5 to 15 in.
Cartridge/Filter PARTICULATE
Certifications/Compliance NIOSH PAPR100-P
Product Type PAPR Healthcare Systems and Kits
Size Medium
Facepiece/Headtop Model Loose fitting HEPA hood
Includes Narrow LF Hood, Shoulder Carriage, Blower Motor, 2 HEPA Filters, 2 Batteries, and Battery Charger
Intrinsic Safety (IS) Rating Yes
Model SALHC20LF2M2
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.