Learn More
Cayman Chemical Salva japonca Salvgen 10mg
SDP
Catalog No. 502913661
Encompass_Preferred
A polyphenol flavonoid with diverse biological activities; inhibits hydrogen peroxide-induced apoptosis and reduces the generation of ROS in SH-SY5Y cells at 25 µM; decreases cleaved caspase-3 levels and the Bax/Bcl-2 ratio in SH-SY5Y cells at 25 and 50 µM; reduces the viability of MCF-7 human breast cancer cells; enhances the DTH response to sheep red blood cells in mice at 3.65-9.68 UG/mouse per day; increases lysate antigen-induced production of IFN-γ in isolated mouse splenocytes; reduces tumor growth in a SMMT model at 9.68 UG/mouse per day
Catalog No. 50-291-3661 Supplier Cayman Chemical Supplier No. 3563210 MG
A polyphenol flavonoid with diverse biological activities; inhibits hydrogen peroxide-induced apoptosis and reduces the generation of ROS in SH-SY5Y cells at 25 µM; decreases cleaved caspase-3 levels and the Bax/Bcl-2 ratio in SH-SY5Y cells at 25 and 50 µM; reduces the viability of MCF-7 human breast cancer cells; enhances the DTH response to sheep red blood cells in mice at 3.65-9.68 UG/mouse per day; increases lysate antigen-induced production of IFN-γ in isolated mouse splenocytes; reduces tumor growth in a SMMT model at 9.68 UG/mouse per day