Learn More
Cayman Chemical Hispidulin, Purity: ≥95%, CAS: 1447-88-7, 10 mg
SDP
Catalog No. 502933393
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits PAF-, arachidonic acid-, and ADP-induced platelet aggregation (IC50s = 20, 4, and 13 μM, respectively); inhibits RANKL-induced osteoclastic differentiation of RAW 264.7 cells and BMMs; inhibits LPS-induced bone resorption in mice at 25 μg/kg; inhibits tumor growth in a Caki-2 mouse xenograft model; reduces sevoflurane-induced cognitive deficits in aged rats at 40 mg/kg; decreases infarct size and brain edema in a rat model of focal cerebral ischemia and reperfusion injury
Catalog No. 50-293-3393 Supplier Cayman Chemical Supplier No. 2638310 MG
A flavonoid with diverse biological activities; inhibits PAF-, arachidonic acid-, and ADP-induced platelet aggregation (IC50s = 20, 4, and 13 μM, respectively); inhibits RANKL-induced osteoclastic differentiation of RAW 264.7 cells and BMMs; inhibits LPS-induced bone resorption in mice at 25 μg/kg; inhibits tumor growth in a Caki-2 mouse xenograft model; reduces sevoflurane-induced cognitive deficits in aged rats at 40 mg/kg; decreases infarct size and brain edema in a rat model of focal cerebral ischemia and reperfusion injury