Learn More
Medchemexpress LLC Salvianolic acid A 5mg
SDP
Catalog No. 5000669437
Encompass_Preferred
Salvianolic acid A (Standard) is the analytical standard of Salvianolic acid A This product is intended for research and analytical applications Salvianolic acid A could protect the blood brain barrier through matrix metallopeptidase 9 (MMP-9) inhibition and anti-inflammation
Catalog No. 50-006-69437 Supplier Medchemexpress LLC Supplier No. 5MGHYN0318R
Add to Cart
Add to Cart
Salvianolic acid A (Standard) is the analytical standard of Salvianolic acid A This product is intended for research and analytical applications Salvianolic acid A could protect the blood brain barrier through matrix metallopeptidase 9 (MMP-9) inhibition and anti-inflammation