Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salvianolic acid A, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015980 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5980 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5980 Supplier MilliporeSigma Supelco Supplier No. 9759910MG
Add to Cart
Add to Cart

Salvianolic acid A is a phenolic acid, isolated from Danshen, a traditional Chinese medicine, commonly used for the treatment of bone loss, chronic renal failure, hepatocirrhosis, hepatitis, coronary heart disease and cerebrovascular disease.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 96574-01-5
Physical Form Neat
Linear Formula C26H22O10
MDL Number MFCD08458447
Synonym (alphaR)-alpha-[[(2 E)-3-[2-[(1 E)-2-(3,4-Dihydroxyphenyl)ethenyl]-3,4-dihydroxyphenyl]-1-oxo-2-propen-1-yl]oxy]-3,4-dihydroxybenzenepropanoic acid
Type Analytical Standard
Quantity 10 mg
Formula Weight 494.45
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.