Learn More
Selleck Chemical LLC Salvianolic acid B S4735-25mg
SDP
Supplier:  Selleck Chemical LLC S473525MG
Encompass_Preferred
Salvianolic acid B (Sal B Lithospermate B Lithospermic acid B) an antioxidant and free radical scavenging compound is the most abundant bioactive compound extracted from the root of Salvia miltiorrhiza Bunge
Catalog No. 50-310-9413