Learn More
Apexbio Technology LLC Salvianolic acid B 115939-25-8; 121521-90-2 20mg
SDP
Catalog No. 5000094375
Encompass_Preferred
Salvianolic acid B is a biologically active polyphenolic compound derived from Salvia miltiorrhiza It modulates cell signaling pathways involved in endothelial function oxidative stress response apoptosis and inflammatory processes Salvianolic acid B exerts its biological activity primarily through antioxidation and anti-inflammatory effects Based on these pharmacological properties Salvianolic acid B holds research potential in cardiovascular protection cerebral ischemia and neurodegenerative disease mechanisms
Catalog No. 50-000-94375 Supplier Apexbio Technology LLC Supplier No. N180620
Add to Cart
Add to Cart
Salvianolic acid B is a biologically active polyphenolic compound derived from Salvia miltiorrhiza It modulates cell signaling pathways involved in endothelial function oxidative stress response apoptosis and inflammatory processes Salvianolic acid B exerts its biological activity primarily through antioxidation and anti-inflammatory effects Based on these pharmacological properties Salvianolic acid B holds research potential in cardiovascular protection cerebral ischemia and neurodegenerative disease mechanisms