Learn More
Medchemexpress LLC HY-N0319 5mg Medchemexpress, Salvianolic acid C CAS:115841-09-3 Purity:>98%
SDP
Catalog No. 501933517
Encompass_Preferred
Medchemexpress, HY-N0319 5mg Salvianolic acid C CAS:115841-09-3 Salvianolic acid C is a noncompetitive Cytochrome P4502C8 ( CYP2C8 ) inhibitor and a moderate mixed inhibitor of Cytochrome P45022J2 ( CYP2J2 ), with K i s of 4.82 μM and 5.75 μM for CYP2C8 and CYP2J2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3517 Supplier Medchemexpress LLC Supplier No. HYN03195MG
Medchemexpress, HY-N0319 5mg Salvianolic acid C CAS:115841-09-3 Salvianolic acid C is a noncompetitive Cytochrome P4502C8 ( CYP2C8 ) inhibitor and a moderate mixed inhibitor of Cytochrome P45022J2 ( CYP2J2 ), with K i s of 4.82 μM and 5.75 μM for CYP2C8 and CYP2J2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.