Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Salvianolic acid E | 142998-46-7 | 1 MG
SDP

Catalog No. 5000417584
Encompass_Preferred

Salvianolic acid E is a natural compound isolated from Salvia miltiorrhiza. It is intended for research use only.

  • High purity (99.83%)
  • Molecular weight: 718.61
  • Molecular formula: C36H30O16
  • Solid appearance
  • Color: Light yellow to yellow
  • Classified as phenylpropanoids and lignans

Catalog No. 50-004-17584 Supplier Medchemexpress LLC Supplier No. HYN75221MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Salvianolic acid E is a natural compound isolated from Salvia miltiorrhiza. It is intended for research use only.

  • High purity (99.83%)
  • Molecular weight: 718.61
  • Molecular formula: C36H30O16
  • Solid appearance
  • Color: Light yellow to yellow
  • Classified as phenylpropanoids and lignans

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.