Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Salvianolic acid F | 158732-59-3 | MFCD31560810 | 99.3% | 314.29 g/mol | C17H14O6 | 5MG
SDP

Catalog No. 5000238353
Encompass_Preferred

Salvianolic acid F is a bioactive phenolic small molecule reported as a KRAS inhibitor, notably against KRAS G12D. It is provided as a solid research reagent with high reported purity for in vitro and preclinical studies, suitable for biochemical and cell-based assays.

  • High reported purity: 99.3%.
  • Solid form suitable for dissolution in common solvents for assay use.
  • Molecular formula C17H14O6; molecular weight 314.29 g/mol.
  • CAS number 158732-59-3 for unambiguous identification.
  • Available in small pack sizes (1 mg-100 mg) for screening and analytical work.

Catalog No. 50-002-38353 Supplier Medchemexpress LLC Supplier No. HY1258475MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Salvianolic acid F is a bioactive phenolic small molecule reported as a KRAS inhibitor, notably against KRAS G12D. It is provided as a solid research reagent with high reported purity for in vitro and preclinical studies, suitable for biochemical and cell-based assays.

  • High reported purity: 99.3%.
  • Solid form suitable for dissolution in common solvents for assay use.
  • Molecular formula C17H14O6; molecular weight 314.29 g/mol.
  • CAS number 158732-59-3 for unambiguous identification.
  • Available in small pack sizes (1 mg-100 mg) for screening and analytical work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.