Learn More
Medchemexpress LLC Salvigenin | 19103-54-9 | MFCD00238678 | 99.9% | 328.32 g·mol⁻¹ | C18H16O6 | 10 MG
SDP
Catalog No. 5000295713
Encompass_Preferred
Salvigenin is a natural polyphenolic flavonoid used in preclinical research for its reported neuroprotective, antitumor, cytotoxic, and immunomodulatory activities. It has been shown to inhibit H2O2-induced apoptosis and is used in biochemical and cell-based studies of ROS signaling and apoptosis pathways. Supplied as a high-purity solid and as a 10 mM solution in DMSO for in vitro use.
  • High purity (≈99.9%) suitable for research applications.
  • Natural polyphenolic flavonoid with neuroprotective and anticancer activities.
  • Inhibits H2O2-induced cell apoptosis; useful for ROS and apoptosis studies.
  • Available as a solid and as a 10 mM solution in DMSO for convenient dosing.
  • Intended for in vitro biochemical and cell-based assays.
Catalog No. 50-002-95713 Supplier Medchemexpress LLC Supplier No. HYN131810MG
Salvigenin is a natural polyphenolic flavonoid used in preclinical research for its reported neuroprotective, antitumor, cytotoxic, and immunomodulatory activities. It has been shown to inhibit H2O2-induced apoptosis and is used in biochemical and cell-based studies of ROS signaling and apoptosis pathways. Supplied as a high-purity solid and as a 10 mM solution in DMSO for in vitro use.
  • High purity (≈99.9%) suitable for research applications.
  • Natural polyphenolic flavonoid with neuroprotective and anticancer activities.
  • Inhibits H2O2-induced cell apoptosis; useful for ROS and apoptosis studies.
  • Available as a solid and as a 10 mM solution in DMSO for convenient dosing.
  • Intended for in vitro biochemical and cell-based assays.