Learn More
Cayman Chemical SalvInolIc AcId A 10mg
SDP
Catalog No. 502979687
Encompass_Preferred
An antioxidant and free radical scavenging compound extracted from S. miltiorrhiza that has been investigated for its cardioprotective and chemopreventative properties
Catalog No. 50-297-9687 Supplier Cayman Chemical Supplier No. 1780010 MG
An antioxidant and free radical scavenging compound extracted from S. miltiorrhiza that has been investigated for its cardioprotective and chemopreventative properties