Learn More
Enzo Life Sciences Salvinorin A (1mg). CAS: 83729-01-5
SDP
Supplier:  Enzo Life Sciences BMLNO1090001
Encompass_Preferred
Potent naturally occuring diterpenoid κ1-opioid selective agonist. Displays high affinity at κ1-opioid receptors (Ki=4.3 nM). Purity: ≥98% (HPLC). Solubility: Soluble in methanol, pyridine, acetonitrile, and chloroform. Long Term Storage: .
Catalog No. 50-201-1200