Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Salvinorin A | 83729-01-5 | MFCD05664742 | 99.0% | 432.46 g/mol | C23H28O8 | 1 MG
SDP

Catalog No. 5000212915
Encompass_Preferred

Salvinorin A is a naturally occurring neoclerodane diterpenoid and a potent, short-acting kappa-opioid receptor (KOPr) agonist used in neuroscience and pharmacology research. Supplied as a high-purity solid, it is intended for receptor binding, functional assays, and analytical reference use.

  • Potent kappa-opioid receptor agonist with high affinity.
  • High reported purity: 98.99% (reported by manufacturer).
  • CAS number 83729-01-5 for unambiguous identification.
  • Formula C23H28O8 and molecular weight 432.46 g/mol.
  • Suitable for receptor binding, functional, and analytical studies.
  • Stored as a powder at -20°C for long-term stability; in solvent, store at -80°C for extended storage.

Catalog No. 50-002-12915 Supplier Medchemexpress LLC Supplier No. HY1008451MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Salvinorin A is a naturally occurring neoclerodane diterpenoid and a potent, short-acting kappa-opioid receptor (KOPr) agonist used in neuroscience and pharmacology research. Supplied as a high-purity solid, it is intended for receptor binding, functional assays, and analytical reference use.

  • Potent kappa-opioid receptor agonist with high affinity.
  • High reported purity: 98.99% (reported by manufacturer).
  • CAS number 83729-01-5 for unambiguous identification.
  • Formula C23H28O8 and molecular weight 432.46 g/mol.
  • Suitable for receptor binding, functional, and analytical studies.
  • Stored as a powder at -20°C for long-term stability; in solvent, store at -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.