Learn More
Cayman Chemical Salvinorin A Carbamate, Purity: ≥90%, CAS: 858345-57-0, 5 mg
SDP
Catalog No. 502987810
Encompass_Preferred
A potent KOR agonist; almost as potent as salvinorin A (EC50 = 6.2 and 4.5 nM for salvinorin A carbamate and salvinorin A, respectively); has increased biological stability compared to salvinorin A
Catalog No. 50-298-7810 Supplier Cayman Chemical Supplier No. 222455 MG
A potent KOR agonist; almost as potent as salvinorin A (EC50 = 6.2 and 4.5 nM for salvinorin A carbamate and salvinorin A, respectively); has increased biological stability compared to salvinorin A